Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hist1h2bg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178196.4 | NP_835503.1 | 1 | 151 | ||
GenBank mRNA | BC060304.1 | - | 1 | 178 | 4217587 |
Gene Symbol: Hist1h2bb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC116853.1 | - | 1 | 179 | 40126172 | |
BC116857.1 | - | 1 | 179 | 40126176 |
Gene Symbol: Hist1h2bc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290380.1 | NP_001277309.1 | 1 | 151 | ||
NM_023422.3 | NP_075911.2 | 1 | 151 | |||
GenBank mRNA | AK005191.1 | - | 1 | 147 | ||
AK005407.1 | - | 1 | 146 | |||
AK011516.1 | - | 1 | 153 | |||
BC019673.1 | - | 1 | 134 | 3993954 | ||
BC082774.1 | - | 1 | 134 |
Gene Symbol: Hist1h2be | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177653.1 | NP_001171124.1 | 2 | 311 | ||
NM_001290530.1 | NP_001277459.1 | 1 | 132 | |||
NM_178194.4 | NP_835501.1 | 2 | 337 | |||
GenBank mRNA | AK030546.1 | - | 2 | 339 | ||
AK049948.1 | - | 2 | 312 | |||
BC069889.1 | - | 1 | 125 | 4217814 |
Gene Symbol: Hist1h2bf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178195.2 | NP_835502.1 | 1 | 118 |
Gene Symbol: Hist1h2bh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178197.2 | NP_835504.1 | 1 | 210 | ||
GenBank mRNA | AK158416.1 | - | 159 | |||
BC092138.1 | - | 125 | 30613720 | |||
BC132584.1 | - | 1 | 210 | 40130861 | ||
BC132586.1 | - | 1 | 210 | 40130863 |
Gene Symbol: Hist1h2bj | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC115792.1 | - | 110 |
Gene Symbol: Hist1h2bk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175665.2 | NP_783596.1 | 1 | 184 | ||
GenBank mRNA | BC147143.1 | - | 1 | 184 | 9056184 | |
BC147144.1 | - | 1 | 184 | 9056197 |
Gene Symbol: Hist1h2bl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178199.2 | NP_835506.1 | 1 | 118 |
Gene Symbol: Hist1h2bm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178200.2 | NP_835507.1 | 1 | 172 | ||
GenBank mRNA | BC139381.1 | - | 1 | 172 | 8862403 | |
BC139382.1 | - | 1 | 172 | 8862404 |
Gene Symbol: Hist1h2bn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178201.2 | NP_835508.1 | 1 | 118 |
Gene Symbol: Hist1h2bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290466.1 | NP_001277395.1 | 1 | 145 | ||
NM_178202.2 | NP_835509.2 | 1 | 145 | |||
GenBank mRNA | AK076671.1 | - | 1 | 146 | ||
BC061044.1 | - | 1 | 145 | 6742370 | ||
M25487.1 | - | 1 | 106 |
Gene Symbol: Hist1h2bq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001097979.2 | NP_001091448.2 | 1 | 146 |
Gene Symbol: Hist1h2br | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110555.1 | NP_001104025.1 | 1 | 146 | ||
GenBank mRNA | AK036389.1 | - | 1 | 147 | ||
BC011440.1 | - | 1 | 131 | 3989862 |