Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stat5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113563.1 | NP_001107035.1 | 2 | 305 | ||
NM_011489.3 | NP_035619.3 | 2 | 637 | |||
GenBank mRNA | AK045531.1 | - | 2 | 447 | ||
AK150098.1 | - | 2 | 274 | |||
AK154014.1 | - | 2 | 637 | |||
AK154664.1 | - | 2 | 271 | |||
AY040231.1 | - | 2 | 591 | |||
AY042906.1 | - | 2 | 591 | |||
AY044901.1 | - | 2 | 591 | |||
AY044902.1 | - | 2 | 591 | |||
AY044903.1 | - | 2 | 591 | |||
BC024319.1 | - | 2 | 261 | 4237132 | ||
U21110.2 | - | 2 | 591 | |||
Z48539.1 | - | 2 | 205 |