Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm3373 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC040090.1 | - | 1922 | 3987869 | ||
BC096548.1 | - | 3224 |
Gene Symbol: 2610042L04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK011748.1 | - | 6 | 1575 |
Gene Symbol: Gm10406 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164727.1 | NP_001158199.1 | 7 | 1735 |
Gene Symbol: Gm10409 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BY608518.1 | - | 1 | 148 |
Gene Symbol: Gm2897 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177715.2 | NP_001171186.1 | 7 | 1762 | ||
GenBank mRNA | BC028273.1 | - | 7 | 1772 |
Gene Symbol: Gm3002 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC080297.1 | - | 5 | 4999 |
Gene Symbol: Gm3020 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK012594.1 | - | 7 | 1774 |
Gene Symbol: Gm3317 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242941.2 | NP_001229870.2 | 5 | 1521 |
Gene Symbol: Gm3383 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291093.1 | NP_001278022.1 | 6 | 1627 | ||
NM_001291094.1 | NP_001278023.1 | 1439 | ||||
GenBank mRNA | BB672657.1 | - | 1 | 192 |
Gene Symbol: Gm3488 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256885.1 | NP_001243814.1 | 5 | 1521 |
Gene Symbol: Gm3500 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256886.1 | NP_001243815.1 | 7 | 1760 |
Gene Symbol: Gm3558 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270842.1 | NP_001257771.1 | 7 | 1835 | ||
GenBank mRNA | BC089473.1 | - | 7 | 1835 | 6588560 |
Gene Symbol: Gm3696 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024712.2 | NP_001019883.2 | 1736 |
Gene Symbol: Gm5796 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029930.2 | NP_001025101.2 | 1595 | |||
GenBank mRNA | BC093494.1 | - | 1595 | 2812218 |
Gene Symbol: LOC100036572 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC027416.1 | - | 2530 |
Gene Symbol: LOC100861615 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270812.1 | NP_001257741.1 | 6 | 1588 | ||
GenBank mRNA | AK049619.1 | - | 6 | 1589 |
Gene Symbol: LOC545005 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC089308.1 | - | 4 | 2315 |