Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145916.2 | NP_666021.2 | 5 | 866 | ||
GenBank mRNA | AK144887.1 | - | 5 | 1078 | ||
AK168948.1 | - | 5 | 821 | |||
BC011501.1 | - | 5 | 884 | 3584546 | ||
BC062103.1 | - | 4 | 2468 |