Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trappc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025432.3 | NP_079708.2 | 3 | 314 | ||
GenBank mRNA | AK002658.1 | - | 4 | 385 | ||
AK004389.1 | - | 339 | ||||
AK007889.1 | - | 3 | 315 | |||
AK008564.1 | - | 4 | 364 | |||
AK034967.1 | - | 3 | 1061 | |||
AK077643.1 | - | 340 | ||||
AY318751.1 | - | 372 | ||||
BC034845.1 | - | 349 | 1528500 | |||
BC061087.1 | - | 384 | 6589013 | |||
BC099551.1 | - | 356 | 30252145 |