Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uqcrq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025352.2 | NP_079628.1 | 2 | 345 | ||
GenBank mRNA | AA510422.1 | - | 2 | 290 | ||
AK002389.1 | - | 3 | 241 | |||
AK003160.1 | - | 3 | 257 | |||
AK005374.1 | - | 327 | ||||
AK007982.1 | - | 2 | 306 | |||
AK203034.1 | - | 2 | 93 | |||
AK214907.1 | - | 1 | 52 | |||
BC027809.1 | - | 1 | 1689 | |||
BC028519.1 | - | 2 | 329 | 1365708 | ||
CB950804.1 | - | 2 | 348 |