Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tshr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113404.1 | NP_001106875.1 | 5 | 510 | ||
NM_011648.5 | NP_035778.3 | 5 | 510 | |||
GenBank mRNA | AK014519.1 | - | 5 | 509 | ||
AK021020.1 | - | 5 | 504 | |||
AK029084.1 | - | 5 | 476 | |||
AK041086.1 | - | 5 | 487 | |||
BB645263.1 | - | 5 | 510 | |||
BC028754.1 | - | 5 | 465 | |||
BC086691.1 | - | 5 | 439 | 30682686 | ||
BC092523.1 | - | 5 | 452 | 30650002 | ||
U02601.1 | - | 5 | 492 | |||
U02602.1 | - | 5 | 492 |