Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcdh18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130448.3 | NP_569715.3 | 1 | 1682 | ||
GenBank mRNA | AF416735.1 | - | 1 | 1627 | ||
AK032820.1 | - | 1 | 1660 | |||
AK036560.1 | - | 1 | 1651 | |||
AK173204.1 | - | 1 | 1627 | |||
BC052198.1 | - | 1 | 1646 | 6512513 |