Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mup7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134675.1 | NP_001128147.1 | 7 | 969 |
Gene Symbol: Mup1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC091744.1 | - | 801 | 5102271 | ||
BC092254.1 | - | 798 | 5100458 | |||
BC100586.1 | - | 818 | 6769126 |
Gene Symbol: Mup15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001200004.1 | NP_001186933.1 | 7 | 825 |