Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tle4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF229633.1 | - | 14, 15 | 1359 | ||
AK122481.2 | - | 12, 13 | 1218 | |||
AK134989.1 | - | 14, 15 | 1993 | |||
BC086631.1 | - | 15, 16 | 1903 | |||
BU149633.1 | - | 397 | ||||
U61363.1 | - | 6, 7 | 762 |