Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnao1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113384.1 | NP_001106855.1 | 4 | 918 | ||
NM_010308.3 | NP_034438.1 | 4 | 918 | |||
GenBank mRNA | AK035907.1 | - | 4 | 919 | ||
AK047197.1 | - | 4 | 922 | |||
BC051989.1 | - | 4 | 745 | 5686124 | ||
M36777.1 | - | 4 | 441 | |||
M36778.1 | - | 4 | 443 |