Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stxbp5l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114611.1 | NP_001108083.1 | 24 | 3191 | ||
NM_001114612.1 | NP_001108084.1 | 25 | 3362 | |||
NM_001114613.1 | NP_001108085.1 | 25 | 3263 | |||
NM_172440.3 | NP_766028.2 | 26 | 3434 | |||
GenBank mRNA | AY542324.1 | - | 25 | 3285 | ||
AY542325.1 | - | 23 | 3042 | |||
AY542326.1 | - | 24 | 3213 | |||
AY542327.1 | - | 24 | 3114 | |||
BU609516.1 | - | 3 | 505 |