Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btaf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080706.1 | NP_001074175.1 | 32 | 4852 | ||
GenBank mRNA | AB287295.1 | - | 32 | 4577 | ||
AK088840.1 | - | 7 | 781 | |||
BC094345.1 | - | 13 | 1969 |