Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CALB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001740.4 | NP_001731.2 | 6, 7 | 566 | ||
NM_007088.3 | NP_009019.1 | 6, 7 | 566 | |||
NR_027910.2 | 6, 7 | 566 | ||||
GenBank mRNA | AK222495.1 | - | 6, 7 | 556 | ||
AK222648.1 | - | 6, 7 | 556 | |||
AK291096.1 | - | 6, 7 | 556 | |||
BC015484.2 | - | 6, 7 | 541 | 3847342 | ||
X56667.1 | - | 6, 7 | 529 |