Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSME4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014614.2 | NP_055429.2 | 14 | 1817 | ||
XM_006711969.1 | XP_006712032.1 | 12 | 1333 | |||
XM_011532705.2 | XP_011531007.1 | 15 | 1841 | |||
XM_011532706.2 | XP_011531008.1 | 15 | 1841 | |||
XM_011532707.2 | XP_011531009.1 | 14 | 1775 | |||
XM_011532709.2 | XP_011531011.1 | 15 | 1841 | |||
GenBank mRNA | AB621805.1 | - | 13 | 1934 | ||
AY894756.1 | - | 14 | 1761 | |||
BC112169.1 | - | 13 | 1453 | 8327637 | ||
BC113668.1 | - | 13 | 1453 | 8322720 | ||
BC143739.1 | - | 13 | 1453 | |||
D38521.1 | - | 14 | 1627 |