Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdzd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081064.1 | NP_001074533.1 | 19 | 6030 | ||
GenBank mRNA | AK122240.1 | - | 1 | 1699 | ||
BC115886.1 | - | 3 | 2083 |