Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSME4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014614.2 | NP_055429.2 | 31, 32 | 3654 | ||
XM_006711969.1 | XP_006712032.1 | 30, 31 | 3231 | |||
XM_011532705.2 | XP_011531007.1 | 32, 33 | 3678 | |||
XM_011532706.2 | XP_011531008.1 | 32, 33 | 3678 | |||
XM_011532707.2 | XP_011531009.1 | 31, 32 | 3612 | |||
GenBank mRNA | AK026085.1 | - | 5, 6 | 651 | ||
AY894754.1 | - | 30, 31 | 3503 | |||
AY894755.1 | - | 12, 13 | 1342 | |||
AY894756.1 | - | 31, 32 | 3598 | |||
BC112169.1 | - | 30, 31 | 3290 | 8327637 | ||
BC113668.1 | - | 30, 31 | 3290 | 8322720 | ||
BC143739.1 | - | 30, 31 | 3290 | |||
D38521.1 | - | 31, 32 | 3464 |