Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm16501 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113395.1 | NP_001106866.1 | 1 | 107 |
Gene Symbol: Gm6026 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177569.1 | NP_001171040.1 | 1 | 107 | ||
GenBank mRNA | EU315689.1 | - | 1 | 107 |