Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tjp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198985.1 | NP_001185914.1 | 7 | 1235 | ||
NM_011597.4 | NP_035727.2 | 7 | 1225 | |||
GenBank mRNA | AF113005.1 | - | 7 | 1083 | ||
AK028494.1 | - | 7 | 1154 | |||
AK217702.1 | - | 2 | 137 | |||
BC034677.1 | - | 7 | 1143 | 3155078 | ||
BC039924.1 | - | 7 | 1154 | 3155079 | ||
BC042695.1 | - | 7 | 1127 |