Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il20rb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033543.3 | NP_001028715.2 | 6, 7 | 958 | ||
GenBank mRNA | AK137306.1 | - | 2, 3 | 810 | ||
BC030430.1 | - | 2, 3 | 266 |