Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033274.3 | NP_001028446.2 | 8 | 2802 | ||
GenBank mRNA | AK220233.1 | - | 7 | 1723 | ||
BC049267.1 | - | 1 | 60 | |||
BC130273.1 | - | 4 | 944 |