Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Terf2ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020584.2 | NP_065609.2 | 3 | 1229 | ||
GenBank mRNA | AK041613.1 | - | 3 | 1175 | ||
AK051818.1 | - | 3 | 1149 | |||
AK081557.1 | - | 3 | 1185 | |||
AK084563.1 | - | 3 | 1177 | |||
AK148508.1 | - | 3 | 1189 | |||
AK148537.1 | - | 3 | 1191 | |||
AK161452.1 | - | 3 | 1208 | |||
AK165074.1 | - | 3 | 1187 | |||
BC012270.1 | - | 3 | 1145 | 3483521 | ||
BC017641.1 | - | 3 | 1205 | 4483320 |