Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clec3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011606.2 | NP_035736.2 | 1, 2 | 213 | ||
GenBank mRNA | BC035043.1 | - | 1, 2 | 165 | 1313548 | |
BY114395.1 | - | 1, 2 | 213 | |||
U08595.1 | - | 1, 2 | 196 | |||
X79199.1 | - | 1, 2 | 188 |