Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110320.1 | NP_001103790.1 | 7, 8 | 1043 | ||
NM_001110321.1 | NP_001103791.1 | 6, 7 | 971 | |||
NM_001110322.1 | NP_001103792.1 | 5, 6 | 860 | |||
NM_007654.2 | NP_031680.2 | 7, 8 | 1022 | |||
GenBank mRNA | AA203771.1 | - | 2, 3 | 196 | ||
AF543214.1 | - | 5, 6 | 719 | |||
AK134132.1 | - | 7, 8 | 1046 | |||
AK144191.1 | - | 6, 7 | 974 | |||
AK151094.1 | - | 862 |