Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLEKHM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015164.2 | NP_055979.2 | 9 | 1708 | ||
XM_005245790.3 | XP_005245847.1 | 8 | 1843 | |||
XM_005245791.4 | XP_005245848.1 | 4 | 976 | |||
XM_017000757.1 | XP_016856246.1 | 9 | 1615 | |||
XM_017000758.1 | XP_016856247.1 | 8 | 1555 | |||
GenBank mRNA | AB020649.1 | - | 9 | 1484 | ||
BC016488.1 | - | 1 | 390 | |||
BC040441.1 | - | 9 | 1711 | |||
BC042103.1 | - | 1 | 38 | |||
BC068599.1 | - | 1 | 63 |