Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BC094916 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024721.2 | NP_001019892.2 | 2 | 431 | ||
GenBank mRNA | AK156095.1 | - | 2 | 427 | ||
AK171847.1 | - | 2 | 422 | |||
BC094916.1 | - | 2 | 325 | 30074914 | ||
JN200820.1 | - | 1 | 125 |