Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rex2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177767.1 | NP_001171238.1 | 5 | 1807 | ||
GenBank mRNA | BC067397.1 | - | 3 | 1562 | 30462971 |
Gene Symbol: Zfp600 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177545.2 | NP_001171016.1 | 5 | 1714 | ||
NM_001177546.2 | NP_001171017.1 | 5 | 1807 | |||
GenBank mRNA | BC108352.1 | - | 5 | 1714 | 30462028 |