Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172704.3 | NP_766292.2 | 14 | 1450 | ||
GenBank mRNA | AK053156.1 | - | 14 | 1432 | ||
AK077544.1 | - | 5 | 384 | |||
AK156793.1 | - | 10 | 4859 | |||
AK171781.1 | - | 14 | 1431 | |||
AK172542.1 | - | 14 | 1431 | |||
AK179457.1 | - | 5 | 366 | |||
AK212087.1 | - | 2 | 98 | |||
BC027574.1 | - | 2 | 100 | |||
BC075619.1 | - | 11 | 1543 | |||
BC079852.1 | - | 14 | 1420 | |||
BC085257.1 | - | 14 | 1424 | 5414963 |