Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ipo8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081113.1 | NP_001074582.1 | 8 | 1064 | ||
GenBank mRNA | AK047854.1 | - | 6 | 634 | ||
AK141137.1 | - | 7 | 741 | |||
BC044819.1 | - | 8 | 1023 | |||
BC054373.1 | - | 8 | 1036 | |||
BC151036.1 | - | 8 | 932 | 9087949 | ||
BC151052.1 | - | 8 | 932 | 9087965 |