Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vmn2r48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105152.1 | NP_001098622.1 | 6 | 1676 |
Gene Symbol: Vmn2r36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105068.1 | NP_001098538.1 | 6 | 1667 |
Gene Symbol: Vmn2r40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105072.1 | NP_001098542.1 | 6 | 1667 |
Gene Symbol: Vmn2r41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105073.1 | NP_001098543.1 | 6 | 1667 |
Gene Symbol: Vmn2r45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105075.1 | NP_001098545.1 | 6 | 1667 |
Set Membership: |
G-protein coupled receptor |