Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnot1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205226.1 | NP_001192155.1 | 35 | 5112 | ||
NM_153164.3 | NP_694804.3 | 35 | 5118 | |||
NM_178078.2 | NP_835179.1 | 35 | 5118 | |||
GenBank mRNA | AK048021.1 | - | 1 | 2121 | ||
AK129258.1 | - | 2078 | ||||
BC018281.1 | - | 8 | 1089 | |||
BC157948.1 | - | 35 | 5031 | 9088123 | ||
BC158073.1 | - | 35 | 5037 | 9007370 | ||
BC172105.1 | - | 35 | 5037 | 9054391 |