Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2200002D01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028179.1 | NP_082455.1 | 4 | 437 | ||
GenBank mRNA | AK008617.1 | - | 4 | 403 | ||
AK033738.1 | - | 362 | ||||
BC086934.1 | - | 4 | 363 |