Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H2-T10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010395.7 | NP_034525.4 | 1243 | |||
GenBank mRNA | AK088241.1 | - | 7 | 745 | ||
AK153876.1 | - | 1232 | ||||
AK161731.1 | - | 7 | 1559 | |||
BC021751.1 | - | 1337 | 4165944 | |||
BC021760.1 | - | 1361 | 5055493 | |||
M35246.1 | - | 9 | 1235 |
Gene Symbol: H2-T9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010399.2 | NP_034529.1 | 1229 | |||
GenBank mRNA | BC044208.1 | - | 1240 | 5036676 | ||
M35248.1 | - | 1229 |