Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rplp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026020.6 | NP_080296.3 | 3 | 210 | ||
GenBank mRNA | AK002419.1 | - | 3 | 177 | ||
AK008344.1 | - | 3 | 211 | |||
AK010618.1 | - | 3 | 217 | |||
AK012402.1 | - | 3 | 205 | |||
AK028140.1 | - | 3 | 209 | |||
AK028149.1 | - | 3 | 211 | |||
AK088737.1 | - | 3 | 210 | |||
AK134206.1 | - | 3 | 210 | |||
AK166774.1 | - | 3 | 175 | |||
BC012413.1 | - | 3 | 192 | 3583752 |
Gene Symbol: Rplp2-ps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC055860.1 | - | 1 | 582 |
Gene Symbol: Wdr89 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028203.1 | NP_082479.1 | 2 | 2407 | ||
GenBank mRNA | AK132326.1 | - | 2 | 2378 |