Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009874.3 | NP_034004.2 | 9 | 779 | ||
GenBank mRNA | AK039084.1 | - | 9 | 767 | ||
AK145759.1 | - | 9 | 705 | |||
AK168286.1 | - | 9 | 778 | |||
BC004605.1 | - | 8 | 626 | 3585145 | ||
BC068160.1 | - | 9 | 764 | 30358255 | ||
BC141043.1 | - | 9 | 736 | 9055655 | ||
U11822.1 | - | 9 | 742 | |||
X74145.1 | - | 9 | 761 |