Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRRC2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015172.3 | NP_055987.2 | 26 | 7384 | ||
XM_005245015.2 | XP_005245072.1 | 26 | 7421 | |||
XM_005245016.4 | XP_005245073.1 | 26 | 7401 | |||
XM_005245017.2 | XP_005245074.1 | 26 | 7421 | |||
XM_005245018.2 | XP_005245075.1 | 26 | 7396 | |||
XM_005245019.2 | XP_005245076.1 | 25 | 7319 | |||
XM_005245020.2 | XP_005245077.1 | 26 | 7421 | |||
XM_005245023.2 | XP_005245080.1 | 26 | 7421 | |||
XM_005245024.2 | XP_005245081.1 | 26 | 7421 | |||
XM_005245025.2 | XP_005245082.1 | 26 | 7421 | |||
XM_005245026.2 | XP_005245083.1 | 26 | 7421 | |||
XM_005245027.2 | XP_005245084.1 | 26 | 7421 | |||
XM_005245028.2 | XP_005245085.1 | 26 | 7421 | |||
XM_005245029.2 | XP_005245086.1 | 26 | 7421 | |||
XM_006711238.3 | XP_006711301.1 | 26 | 7401 | |||
XM_017000760.1 | XP_016856249.1 | 26 | 7386 | |||
XM_017000761.1 | XP_016856250.1 | 26 | 7421 | |||
XM_017000762.1 | XP_016856251.1 | 25 | 7319 | |||
GenBank mRNA | AB029019.2 | - | 12 | 4189 | ||
AF488829.1 | - | 25 | 7118 | |||
AK000190.1 | - | 7 | 1035 | |||
AK022492.1 | - | 5 | 708 | |||
AK304841.1 | - | 5 | 735 | |||
AL096857.1 | - | 25 | 7175 | |||
AL162004.1 | - | 9 | 2566 |