Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mdh1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029696.4 | NP_083972.3 | 6 | 988 | ||
GenBank mRNA | AK007262.1 | - | 2 | 113 | ||
BC050786.1 | - | 6 | 988 | 6703279 |