Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgef11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003912.1 | NP_001003912.1 | 3 | 191 | ||
GenBank mRNA | AK157225.1 | - | 3 | 1372 | ||
AK165441.1 | - | 3 | 1351 | |||
AK171406.1 | - | 3 | 1386 | |||
BC072663.1 | - | 3 | 841 | |||
BC079565.1 | - | 3 | 191 | 30536257 | ||
BC079915.1 | - | 3 | 689 |