Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TBC1D1 SIRNA Assays ›
Gene Symbol:
TBC1D1
Gene Name:
TBC1 domain family member 1
Gene Aliases:
TBC, TBC1
Chromosome Location:
Chr.4: 37891084 - 38139175 on Build GRCh38
UniGene ID:
Hs.176503
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s23309
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TBC1D1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001253912.1
NP_001240841.1
18
3250
NM_001253913.1
NP_001240842.1
2
607
NM_001253914.1
NP_001240843.1
2
253
NM_015173.3
NP_055988.2
16
2968
XM_005262646.2
XP_005262703.1
19
3289
XM_005262647.2
XP_005262704.1
18
3250
XM_005262649.2
XP_005262706.1
18
2742
XM_006714001.2
XP_006714064.1
7
1069
XM_011513659.2
XP_011511961.1
20
3349
XM_011513660.2
XP_011511962.1
19
3310
XM_011513662.2
XP_011511964.1
19
3229
XM_011513663.2
XP_011511965.1
18
3190
XM_011513664.2
XP_011511966.1
19
3187
XM_011513665.2
XP_011511967.1
18
3067
XM_011513666.2
XP_011511968.1
17
3028
XM_011513670.2
XP_011511972.1
5
1071
XM_017007918.1
XP_016863407.1
18
3148
XM_017007919.1
XP_016863408.1
17
3130
XM_017007920.1
XP_016863409.1
17
3007
XM_017007921.1
XP_016863410.1
15
2283
GenBank mRNA
AB029031.1
-
10
1395
AB449881.1
-
15
2610
AK027355.1
-
8
1167
AK057182.1
-
2
607
AK074954.1
-
2
253
AK295746.1
-
18
3073
BC014529.2
-
14
1806
BC050321.1
-
16
2965
5262043
BC053648.1
-
14
1781
BC126979.1
-
6
1083
Back To Top
More Information
Panther Classification:
Molecular Function -
scaffold/adaptor protein
GTPase-activating protein
Gene Ontology Categories:
Function(s)
Process(es)
intracellular protein transport
regulation of vesicle fusion
regulation of protein localization
membrane organization
activation of GTPase activity
regulation of cilium assembly
GTPase activator activity
protein binding
Rab GTPase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-hnnbr:80/10.239.164.27:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline