Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ston1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029858.2 | NP_084134.2 | 2 | 2032 | ||
GenBank mRNA | AK014958.1 | - | 2 | 1967 | ||
AK029878.1 | - | 1979 | ||||
AK029959.1 | - | 2012 | ||||
AK077966.1 | - | 2 | 1760 | |||
AK136565.1 | - | 2 | 1886 | |||
CF729405.1 | - | 1 | 236 |