Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015174.1 | NP_055989.1 | 2, 3 | 284 | ||
XM_011527831.2 | XP_011526133.1 | 2, 3 | 283 | |||
GenBank mRNA | AB029009.1 | - | 1, 2 | 162 | ||
AK295037.1 | - | 2, 3 | 283 | |||
DA333602.1 | - | 2, 3 | 284 |