Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Esco2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028039.2 | NP_082315.3 | 10 | 1779 | ||
GenBank mRNA | AA881675.1 | - | 2 | 261 | ||
AK010391.1 | - | 5 | 604 | |||
AK144685.1 | - | 10 | 1735 | |||
BC033303.1 | - | 10 | 1694 | 3675568 | ||
BC071275.1 | - | 10 | 1712 | 6331551 |