Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Idua | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008325.4 | NP_032351.2 | 5, 6 | 913 | ||
NM_172997.1 | NP_766585.1 | 4, 5 | 772 | |||
GenBank mRNA | AK031414.1 | - | 4, 5 | 567 | ||
AK045330.1 | - | 3, 4 | 400 | |||
AK155627.1 | - | 5, 6 | 914 | |||
BC057935.1 | - | 4, 5 | 534 | 5327150 | ||
L34111.1 | - | 4, 5 | 487 |