Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nr1i2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098404.1 | NP_001091874.1 | 6 | 1084 | ||
NM_010936.3 | NP_035066.1 | 6 | 1207 | |||
GenBank mRNA | AF031814.1 | - | 6 | 1022 | ||
AK018630.1 | - | 6 | 1207 | |||
BC120796.1 | - | 6 | 954 | 40129719 | ||
BC137780.1 | - | 6 | 954 | 8860799 |