Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hist2h2aa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK002725.1 | - | 1 | 525 | ||
AK006728.1 | - | 1 | 524 | |||
BC010564.1 | - | 1 | 508 | 3582122 | ||
BC053966.1 | - | 1 | 517 | |||
BC062255.1 | - | 1 | 508 | 1448126 | ||
BC064002.1 | - | 634 | ||||
BC089519.1 | - | 1 | 565 | 6771160 | ||
Z30940.1 | - | 1 | 503 |