Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198859.1 | NP_001185788.1 | 8 | 1226 | ||
NM_001198860.1 | NP_001185789.1 | 9 | 1419 | |||
NM_001198861.1 | NP_001185790.1 | 9 | 1419 | |||
NM_013502.3 | NP_038530.1 | 8 | 1226 | |||
GenBank mRNA | AB033122.1 | - | 8 | 1191 | ||
AJ010483.1 | - | 8 | 1139 | |||
AK133816.1 | - | 9 | 1390 | |||
AK149906.1 | - | 7 | 2413 | |||
AK160658.1 | - | 9 | 1409 | |||
AK165276.1 | - | 9 | 1374 | |||
AK170133.1 | - | 7 | 933 | |||
AK171650.1 | - | 7 | 1059 | |||
BC013702.1 | - | 8 | 1139 | 3155102 | ||
BC015071.1 | - | 8 | 1195 | 4192052 | ||
BC042425.1 | - | 9 | 1400 | 5005681 |