Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ric1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081319.1 | NP_001074788.1 | 18 | 2251 | ||
GenBank mRNA | AK173171.1 | - | 17 | 1833 | ||
BC036137.1 | - | 2 | 47 |