Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lyz2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017372.3 | NP_059068.1 | 2 | 247 | ||
GenBank mRNA | AK148516.1 | - | 2 | 248 | ||
AK150998.1 | - | 2 | 250 | |||
AK153244.1 | - | 2 | 250 | |||
AK153475.1 | - | 2 | 248 | |||
AK159276.1 | - | 2 | 297 | |||
AK159640.1 | - | 2 | 244 | |||
BC002069.1 | - | 2 | 239 | 3593360 | ||
BC054463.1 | - | 2 | 239 | 6514977 |