Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010911.2 | NP_035041.2 | 6, 7 | 717 | ||
GenBank mRNA | AK005724.1 | - | 6, 7 | 706 | ||
AK075575.1 | - | 6, 7 | 715 | |||
AK132838.1 | - | 6, 7 | 718 | |||
AK148527.1 | - | 6, 7 | 718 | |||
AK159828.1 | - | 6, 7 | 706 | |||
AK183689.1 | - | 2, 3 | 123 | |||
AK206231.1 | - | 2, 3 | 123 | |||
AK206381.1 | - | 2, 3 | 123 | |||
AK206572.1 | - | 2, 3 | 123 | |||
AK207148.1 | - | 2, 3 | 123 | |||
AK207847.1 | - | 2, 3 | 123 | |||
AK207895.1 | - | 2, 3 | 123 | |||
AK211411.1 | - | 2, 3 | 123 | |||
AK211628.1 | - | 2, 3 | 123 | |||
AK212905.1 | - | 2, 3 | 123 | |||
AK216163.1 | - | 1, 2 | 123 | |||
AK220082.1 | - | 1, 2 | 123 | |||
BC010586.1 | - | 6, 7 | 695 | 3711352 |