Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ercc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011729.2 | NP_035859.2 | 11 | 2779 | ||
GenBank mRNA | AK137563.1 | - | 12 | 2611 | ||
AK140825.1 | - | 11 | 2608 | |||
AK160581.1 | - | 5 | 1653 | |||
AK160645.1 | - | 9 | 2151 | |||
AK164205.1 | - | 11 | 2597 | |||
BC033413.1 | - | 11 | 2499 | |||
BC053034.1 | - | 11 | 2589 | |||
BC060659.1 | - | 10 | 2910 | |||
BC092235.1 | - | 11 | 2570 | |||
BC141070.1 | - | 11 | 2609 | 9055682 | ||
D16306.1 | - | 11 | 2474 | |||
U40796.1 | - | 11 | 2675 |